Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383222.1 | 18 - 19 | 2485 | 137 | ||
NM_070018.6 | NP_502419.2 | 13 - 14 | 1753 | 137 | ||
GenBank mRNA | AB055112.1 | - | 15 - 16 | 2150 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |