Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383222.1 | 22 - 23 | 3114 | 76 | ||
NM_070018.6 | NP_502419.2 | 17 - 18 | 2382 | 76 | ||
GenBank mRNA | AB055112.1 | - | 19 - 20 | 2779 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |