Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383222.1 | 6 - 7 | 981 | 69 | ||
NM_070018.6 | NP_502419.2 | 1 - 2 | 249 | 69 | ||
GenBank mRNA | AB055112.1 | - | 3 - 4 | 646 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |