Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268735.4 | NP_001255664.1 | 6 - 7 | 1213 | 69 | |
NM_001268737.4 | NP_001255666.1 | 7 - 8 | 1268 | 69 | ||
NM_001268738.3 | NP_001255667.1 | 6 - 7 | 1237 | 69 | ||
NM_001306974.3 | NP_001293903.1 | 7 - 8 | 1249 | 69 | ||
NM_001306975.3 | NP_001293904.1 | 7 - 8 | 1249 | 69 | ||
NM_001306979.3 | NP_001293908.1 | 6 - 7 | 1213 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |