Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026105.4 | NP_001021276.1 | 1 - 2 | 290 | 73 | |
NM_001026106.2 | NP_001021277.1 | 1 - 2 | 177 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |