Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028095.6 | NP_001023266.1 | 6 - 7 | 1521 | 109 | |
GenBank mRNA | AF283322.1 | - | 6 - 7 | 1541 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |