Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383230.1 | 5 - 6 | 1136 | 58 | ||
NM_070161.5 | NP_502562.1 | 5 - 6 | 1178 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |