Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsd-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047487.3 | NP_001040952.2 | 14 - 15 | 2585 | 88 | |
NM_001047488.4 | NP_001040953.1 | 11 - 12 | 2015 | 88 | ||
NM_001268820.3 | NP_001255749.1 | 7 - 8 | 1241 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |