Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070242.2 | NP_502643.1 | 4 - 5 | 856 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |