Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tlp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070246.5 | NP_502647.1 | 2 - 3 | 719 | 68 | |
GenBank mRNA | AY046083.1 | - | 2 - 3 | 617 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |