Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sas-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070259.6 | NP_502660.1 | 1 - 2 | 102 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |