Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268846.3 | NP_001255775.1 | 3 - 4 | 370 | 87 | |
NM_001268847.4 | NP_001255776.1 | 2 - 3 | 219 | 87 | ||
NM_001307070.3 | NP_001293999.1 | 2 - 3 | 192 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |