Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nkcc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083255.3 | NP_001076724.1 | 2 - 3 | 290 | 77 | |
NM_001268861.4 | NP_001255790.1 | 3 - 4 | 354 | 77 | ||
NM_001268862.4 | NP_001255791.1 | 2 - 3 | 262 | 77 | ||
NM_001276883.3 | NP_001263812.1 | 2 - 3 | 261 | 77 | ||
NM_070303.2 | NP_502704.1 | 2 - 3 | 290 | 77 | ||
NM_070304.1 | NP_502705.1 | 2 - 3 | 261 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |