Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nkcc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083255.3 | NP_001076724.1 | 8 - 9 | 2346 | 117 | |
NM_001268859.3 | NP_001255788.1 | 4 - 5 | 1297 | 117 | ||
NM_001268860.3 | NP_001255789.1 | 1 - 2 | 331 | 117 | ||
NM_001268861.4 | NP_001255790.1 | 9 - 10 | 2410 | 117 | ||
NM_001268862.4 | NP_001255791.1 | 8 - 9 | 2318 | 117 | ||
NM_001276883.3 | NP_001263812.1 | 8 - 9 | 2317 | 117 | ||
NM_070303.2 | NP_502704.1 | 8 - 9 | 2346 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |