Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070309.3 | NP_502710.2 | 6 - 7 | 1711 | 130 | |
NR_131638.1 | 6 - 7 | 1379 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |