Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sqrd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268869.4 | NP_001255798.1 | 2 - 3 | 528 | 60 | |
NM_001268870.3 | NP_001255799.1 | 2 - 3 | 539 | 60 | ||
NM_001268871.3 | NP_001255800.1 | 1 - 2 | 183 | 60 | ||
NM_001307012.3 | NP_001293941.1 | 1 - 2 | 183 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |