Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sqrd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268869.4 | NP_001255798.1 | 7 - 8 | 1336 | 71 | |
NM_001268870.3 | NP_001255799.1 | 7 - 8 | 1200 | 71 | ||
NM_001268871.3 | NP_001255800.1 | 6 - 7 | 844 | 71 | ||
NM_001307012.3 | NP_001293941.1 | 6 - 7 | 991 | 71 | ||
NM_001307302.3 | NP_001294231.1 | 4 - 5 | 445 | 71 | ||
NM_001307303.3 | NP_001294232.1 | 4 - 5 | 592 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |