Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eps-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047582.5 | NP_001041047.1 | 2 - 3 | 261 | 79 | |
NM_001047583.7 | NP_001041048.1 | 2 - 3 | 267 | 79 | ||
NM_001306998.2 | NP_001293927.1 | 2 - 3 | 260 | 79 | ||
NM_001372963.2 | NP_001360610.1 | 2 | 188 | 79 | ||
NM_001372964.2 | NP_001360611.1 | 2 | 188 | 79 | ||
GenBank mRNA | AY611484.1 | - | 2 - 3 | 185 | 79 | |
AY611485.1 | - | 2 - 3 | 185 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |