Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070439.4 | NP_502840.4 | 1 - 2 | 182 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |