Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070443.3 | NP_502844.1 | 1 - 2 | 55 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |