Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307187.2 | NP_001294116.1 | 13 - 14 | 2275 | 121 | |
NM_001307188.3 | NP_001294117.1 | 12 - 13 | 2089 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |