Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307187.2 | NP_001294116.1 | 15 - 16 | 2553 | 100 | |
NM_001307188.3 | NP_001294117.1 | 14 - 15 | 2367 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |