Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070492.7 | NP_502893.2 | 1 - 2 | 208 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |