Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070492.7 | NP_502893.2 | 2 - 3 | 412 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |