Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047546.3 | NP_001041011.1 | 3 - 4 | 427 | 86 | |
NM_001047547.3 | NP_001041012.1 | 1 - 2 | 67 | 86 | ||
NM_001307189.3 | NP_001294118.1 | 3 - 4 | 427 | 86 | ||
NM_001307190.3 | NP_001294119.1 | 1 - 2 | 67 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |