Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jac-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047542.3 | NP_001041007.1 | 5 - 6 | 800 | 76 | |
NM_001047543.4 | NP_001041008.1 | 2 - 3 | 290 | 76 | ||
NM_001268952.3 | NP_001255881.1 | 3 - 4 | 581 | 76 | ||
NM_001307194.3 | NP_001294123.1 | 5 - 6 | 800 | 76 | ||
NM_001307195.3 | NP_001294124.1 | 3 - 4 | 581 | 76 | ||
NM_001307196.3 | NP_001294125.1 | 2 - 3 | 290 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |