Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gln-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047544.7 | NP_001041009.1 | 2 - 3 | 344 | 74 | |
NM_001047545.4 | NP_001041010.1 | 2 - 3 | 335 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |