Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sta-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129342.4 | NP_001122814.1 | 4 - 5 | 1015 | 99 | |
NM_001129343.3 | NP_001122815.1 | 4 - 5 | 1017 | 99 | ||
NM_070573.6 | NP_502974.2 | 4 - 5 | 1017 | 99 | ||
GenBank mRNA | AY943817.1 | - | 4 - 5 | 1039 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |