Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mel-46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268980.3 | NP_001255909.1 | 3 - 4 | 433 | 69 | |
NM_001372971.2 | NP_001360155.1 | 3 - 4 | 433 | 69 | ||
GenBank mRNA | EU051652.1 | - | 3 - 4 | 449 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |