Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atgp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380570.1 | NP_001367158.1 | 2 - 3 | 419 | 76 | |
NM_070663.7 | NP_503064.2 | 9 - 10 | 1728 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |