Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026175.4 | NP_001021346.1 | 15 - 16 | 3681 | 69 | |
NM_001372980.2 | NP_001360389.1 | 15 | 3369 | 69 | ||
NM_001372988.2 | NP_001360397.1 | 13 | 3105 | 69 | ||
NM_001380576.2 | NP_001368552.1 | 16 - 17 | 3742 | 69 | ||
NM_001380577.1 | NP_001368553.1 | 15 - 16 | 3612 | 69 | ||
NM_001380578.2 | NP_001368554.1 | 14 - 15 | 3430 | 69 | ||
NM_001380579.1 | NP_001368555.1 | 13 - 14 | 3189 | 69 | ||
NM_001380580.2 | NP_001368556.1 | 15 - 16 | 3842 | 69 | ||
NM_001380581.2 | NP_001368557.1 | 14 - 15 | 3603 | 69 | ||
NM_001380582.2 | NP_001368558.1 | 14 - 15 | 3510 | 69 | ||
NM_001380583.1 | NP_001368559.1 | 13 - 14 | 3260 | 69 | ||
NM_001380584.1 | NP_001368560.1 | 13 - 14 | 3160 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |