Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068605.7 | NP_501006.1 | 4 - 5 | 1031 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |