Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068617.8 | NP_501018.1 | 3 - 4 | 335 | 151 | |
GenBank mRNA | AF480439.1 | - | 3 - 4 | 330 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |