Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyci-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372852.3 | NP_001360494.1 | 3 | 466 | 58 | |
NM_001383110.1 | 2 - 3 | 367 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |