Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tre-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306844.3 | NP_001293773.1 | 5 - 6 | 539 | 109 | |
NM_001380293.1 | NP_001368400.1 | 4 - 5 | 398 | 109 | ||
NM_068657.7 | NP_501058.2 | 7 - 8 | 797 | 109 | ||
GenBank mRNA | AJ512334.1 | - | 6 - 7 | 774 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |