Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380302.1 | NP_001367449.1 | 2 - 3 | 201 | 123 | |
NM_001380303.1 | NP_001367448.1 | 2 - 3 | 182 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |