Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380302.1 | NP_001367449.1 | 7 - 8 | 857 | 108 | |
NM_001380303.1 | NP_001367448.1 | 7 - 8 | 838 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |