Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313231.3 | NP_001300160.1 | 12 - 13 | 2057 | 98 | |
NM_001313232.3 | NP_001300161.1 | 12 - 13 | 1922 | 98 | ||
NM_001313233.3 | NP_001300162.1 | 1 - 2 | 173 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |