Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070802.1 | NP_503203.1 | 1 - 2 | 175 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |