Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070814.5 | NP_503215.2 | 2 - 3 | 286 | 79 | |
GenBank mRNA | AY204171.1 | - | 2 - 3 | 295 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |