Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sri-73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070846.4 | NP_503247.2 | 2 - 3 | 285 | 86 | |
NR_132479.1 | 2 - 3 | 285 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |