Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070898.1 | NP_503299.1 | 3 - 4 | 452 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |