Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fmo-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070951.5 | NP_503352.1 | 4 - 5 | 831 | 143 | |
GenBank mRNA | AJ582073.1 | - | 4 - 5 | 745 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |