Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sqv-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313350.3 | NP_001300279.1 | 2 - 3 | 825 | 63 | |
NM_070958.6 | NP_503359.2 | 5 - 6 | 1971 | 63 | ||
GenBank mRNA | AJ496235.1 | - | 5 - 6 | 1959 | 63 | |
AY241928.1 | - | 5 - 6 | 1982 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |