Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029083.5 | NP_001024254.1 | 2 - 3 | 441 | 70 | |
NM_001313344.4 | NP_001300273.1 | 2 | 483 | 70 | ||
NM_001313345.3 | NP_001300274.1 | 2 - 3 | 453 | 70 | ||
NM_001313346.3 | NP_001300275.1 | 1 - 2 | 60 | 70 | ||
NM_070959.8 | NP_503360.4 | 2 - 3 | 483 | 70 | ||
GenBank mRNA | AF535160.1 | - | 3 - 4 | 678 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |