Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ergo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070961.9 | NP_503362.2 | 2 - 3 | 1134 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |