Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380597.1 | NP_001368562.1 | 13 - 14 | 2385 | 82 | |
NM_071001.5 | NP_503402.3 | 13 - 14 | 2500 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |