Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380597.1 | NP_001368562.1 | 3 - 4 | 312 | 97 | |
NM_071001.5 | NP_503402.3 | 3 - 4 | 436 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |