Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snt-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313247.3 | NP_001300176.1 | 2 - 3 | 102 | 58 | |
NM_071008.5 | NP_503409.1 | 2 - 3 | 198 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |