Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028901.3 | NP_001024072.1 | 2 - 3 | 242 | 81 | |
NM_001028902.1 | NP_001024073.1 | 2 - 3 | 242 | 81 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |